Search
Search

Mir6960| Gene Symbol: Mir6960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105925.1 | 1 | 31 | 59 | ||
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 34284 |
Gm34284 | -6.1kb | Anti-sense | Chr. 15 - 99068138 - 99074669 on NCBI Build GRCm38 | |
| Downstream |
complement component 1, q subcomponent-like 4 |
C1ql4 |
C1qtnf11, CTRP11 |
3.2kb | Anti-sense | Chr. 15 - 99084086 - 99090694 on NCBI Build GRCm38 |
Set Membership: |
|