Search
Search

6820408C15Rik| Gene Symbol: 6820408C15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110367.1 | 10 - 11 | 1347 | 87 | ||
| NR_110368.1 | 9 - 10 | 1247 | 87 | |||
| GenBank mRNA | BC144943.1 | - | 9 - 10 | 1224 | 87 | |
| BC144945.1 | - | 8 - 9 | 1120 | 87 | ||
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
zinc finger, CCHC domain containing 3 |
Zcchc3 | 2810406K24Rik | -26.2kb | Anti-sense | Chr. 2 - 152411956 - 152415044 on NCBI Build GRCm38 |
| Downstream | defensin beta 23 | Defb23 | EG629114 | 16.6kb | Anti-sense | Chr. 2 - 152459055 - 152464620 on NCBI Build GRCm38 |
Set Membership: |
|