Search
Search

Pdcd5| Gene Symbol: Pdcd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019746.4 | NP_062720.1 | 2 - 3 | 220 | 72 | |
| GenBank mRNA | AF161074.1 | - | 2 - 3 | 109 | 72 | |
| AK007297.1 | - | 2 - 3 | 133 | 72 | ||
| AK008632.1 | - | 2 - 3 | 234 | 72 | ||
| BC048476.1 | - | 2 - 3 | 120 | 6773378 | 72 | |
| BC056167.1 | - | 2 - 3 | 112 | 5712632 | 72 | |
| BC092092.1 | - | 2 - 3 | 3492310 | 72 | ||
Set Membership: |
|