Search
Search

Zfp11| Gene Symbol: Zfp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172462.4 | NP_766050.1 | 3 | 1782 | 76 | |
| GenBank mRNA | AK032872.1 | - | 3 | 1797 | 76 | |
| AK085077.1 | - | 2 | 1768 | 76 | ||
| BC058553.1 | - | 3 | 1780 | 76 | ||
| BC094590.1 | - | 2 | 1767 | 30616053 | 76 | |
| BC139312.1 | - | 2 | 1762 | 8862334 | 76 | |
| BC139315.1 | - | 2 | 1762 | 8862337 | 76 | |
Set Membership: |
|