Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm05724508_g1
          See other GAPDH GE Assays ›
          Gene Symbol:
          Gapdh Gm12070 Gm5523
          Gene Name:
          glyceraldehyde-3-phosphate dehydrogenase
          glyceraldehyde-3-phosphate dehydrogenase pseudogene
          glyceraldehyde-3-phosphate dehydrogenase pseudogene
          Gene Aliases:
          Gapd
          OTTMUSG00000005300
          EG433273
          Chromosome Location:
          Chr.6: 125161338 - 125166511 on Build GRCm38
          Chromosome Location:
          Chr.11: 26785108 - 26787860 on Build GRCm38
          Chromosome Location:
          Chr.1: 13785685 - 13786929 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          Assay ID Mm05724508_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gapdh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001289726.1 NP_001276655.1 4 513 80
          NM_008084.3 NP_032110.1 4 661 80
          GenBank mRNA AK002273.1 - 4 494 80
          AK081405.1 - 4 494 80
          AK140794.1 - 4 494 80
          AK144690.1 - 4 515 80
          AK146435.1 - 4 494 80
          AK147738.1 - 4 494 80
          AK147891.1 - 4 494 80
          AK160399.1 - 4 494 80
          AK164415.1 - 4 492 80
          AK168217.1 - 4 494 80
          AK169742.1 - 1 2064 80
          AK178680.1 - 1 62 80
          AK184690.1 - 1 71 80
          AK197268.1 - 1 62 80
          AK200636.1 - 1 62 80
          AK201486.1 - 1 62 80
          AK204478.1 - 1 63 80
          BC020407.1 - 4 482 80
          BC023196.1 - 4 492 80
          BC082592.1 - 4 471 30634381 80
          BC083065.1 - 4 470 5371133 80
          BC083079.1 - 4 476 6493407 80
          BC083080.1 - 4 478 6494145 80
          BC083149.1 - 457 30459585 80
          BC085274.1 - 479 6530979 80
          BC085275.1 - 4 488 30444086 80
          BC091768.1 - 457 4159824 80
          BC092252.1 - 4 480 5044783 80
          BC092264.1 - 482 2655526 80
          BC092267.1 - 468 4010884 80
          BC092294.1 - 471 5068049 80
          BC093508.1 - 451 4975394 80
          BC094037.1 - 458 5009636 80
          BC095932.1 - 4 482 30733532 80
          BC096440.1 - 4 482 2649847 80
          BC096590.1 - 482 6486174 80
          BC110311.1 - 4 483 5340298 80
          BC145810.1 - 4 464 40131113 80
          BC145812.1 - 4 464 40131115 80
          BU504528.1 - 479 80
          DQ403054.1 - 2 352 80
          GU214026.1 - 4 492 80
          M32599.1 - 4 467 80
          Gene Symbol: Gm12070
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002890.1 2 840 80
          GenBank mRNA BC098095.1 - 2 840 80
          Gene Symbol: Gm5523
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_004447.1 1 466 80
          GenBank mRNA AK188672.1 - 71 80
          AK193983.1 - 62 80

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          carbohydrate metabolic process
          gluconeogenesis
          glycolytic process
          apoptotic process
          multicellular organism development
          negative regulation of translation
          peptidyl-cysteine S-trans-nitrosylation
          protein stabilization
          neuron apoptotic process
          oxidation-reduction process
          response to ammonium ion
          cellular response to interferon-gamma
          glyceraldehyde-3-phosphate dehydrogenase (NAD+) (phosphorylating) activity
          microtubule binding
          peptidyl-cysteine S-nitrosylase activity
          identical protein binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline