Search
Search

Thrap3| Gene Symbol: Thrap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146153.3 | NP_666265.3 | 4 - 5 | 1889 | 99 | |
| GenBank mRNA | AK034044.1 | - | 4 - 5 | 1769 | 99 | |
| BC012655.1 | - | 1 - 2 | 575 | 99 | ||
| BC040346.1 | - | 4 - 5 | 1820 | 99 | ||
| BC092239.1 | - | 4 - 5 | 1805 | 30659321 | 99 | |
Set Membership: |
|