Search
Search

Armcx6| Gene Symbol: Armcx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007578.2 | NP_001007579.1 | 4 | 1240 | 93 | |
| GenBank mRNA | AK148507.1 | - | 4 | 1240 | 93 | |
| BC027264.1 | - | 4 | 1233 | 3492516 | 93 | |
Set Membership: |
|