Search
Search

1700030C10Rik| Gene Symbol: 1700030C10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015521.1 | 2 | 928 | 76 | ||
| GenBank mRNA | AK006529.1 | - | 2 | 716 | 76 | |
| BC100365.1 | - | 2 | 928 | 76 | ||
Set Membership: |
|