Search
Search

Cryaa| Gene Symbol: Cryaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278569.1 | NP_001265498.1 | 4 | 1091 | 101 | |
| NM_001278570.1 | NP_001265499.1 | 3 | 1004 | 101 | ||
| NM_013501.3 | NP_038529.1 | 4 | 1073 | 101 | ||
| GenBank mRNA | AK143463.1 | - | 3 | 1003 | 101 | |
| BC085170.1 | - | 4 | 1059 | 30699322 | 101 | |
| BC085172.1 | - | 3 | 990 | 30699371 | 101 | |
| BC092385.1 | - | 3 | 945 | 30602480 | 101 | |
| CA452236.1 | - | 2 | 654 | 101 | ||
| J00376.1 | - | 3 | 910 | 107 | ||
Set Membership: |
|