Search
Search
| Gene Symbol: Aptx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025444.3 | NP_001020615.1 | 8 | 995 | 79 | |
| NM_001025445.2 | NP_001020616.1 | 8 | 990 | 79 | ||
| NM_025545.4 | NP_079821.3 | 8 | 1130 | 79 | ||
| GenBank mRNA | AK005286.1 | - | 7 | 947 | 79 | |
| AK010516.1 | - | 8 | 977 | 79 | ||
| AK017522.1 | - | 7 | 958 | 79 | ||
| AK077351.1 | - | 8 | 1131 | 79 | ||
| AK088928.1 | - | 7 | 680 | 79 | ||
| AY040780.1 | - | 8 | 1141 | 79 | ||
| AY208844.1 | - | 8 | 1142 | 79 | ||
| BC021872.1 | - | 8 | 988 | 3994375 | 79 | |
| BC068309.1 | - | 7 | 903 | 79 | ||
| BC071213.1 | - | 8 | 982 | 79 | ||
Set Membership: |
|