Search
Search

Cend1| Gene Symbol: Cend1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021316.4 | NP_067291.1 | 2 | 1142 | 88 | |
| GenBank mRNA | AK005092.1 | - | 2 | 1117 | 88 | |
| AK079045.1 | - | 2 | 1143 | 88 | ||
| BC023032.1 | - | 2 | 1080 | 5366891 | 88 | |
Set Membership: |
|