Search
Search

| Gene Symbol: Wdr59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170742.1 | NP_001164213.1 | 26 | 3632 | 70 | |
| NM_001170743.1 | NP_001164214.1 | 27 | 3689 | 70 | ||
| NM_176923.4 | NP_795897.2 | 27 | 3683 | 70 | ||
| GenBank mRNA | AK030644.1 | - | 27 | 3564 | 70 | |
| AK052262.1 | - | 3 | 1038 | 70 | ||
| AK136303.1 | - | 22 | 3184 | 70 | ||
| AK170910.1 | - | 26 | 3631 | 70 | ||
| AK173301.1 | - | 27 | 3652 | 70 | ||
| BC066082.1 | - | 26 | 3617 | 30355918 | 70 | |
| EF011622.1 | - | 26 | 3617 | 70 | ||
| Gene Symbol: Mir1957b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105819.1 | 1 | 70 | |||
Set Membership: |
|