Search
Search

Degs2| Gene Symbol: Degs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171002.1 | NP_001164473.1 | 1 - 2 | 150 | 64 | |
| NM_027299.5 | NP_081575.2 | 1 - 2 | 150 | 64 | ||
| GenBank mRNA | AF466377.1 | - | 1 - 2 | 138 | 64 | |
| AK008666.1 | - | 1 - 2 | 151 | 64 | ||
| AK162979.1 | - | 1 - 2 | 143 | 64 | ||
| BC016427.1 | - | 1 - 2 | 132 | 4218832 | 64 | |
| BC035454.1 | - | 1 - 2 | 132 | 64 | ||
Set Membership: |
|