Search
Search

A630076J17Rik| Gene Symbol: A630076J17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256174.1 | NP_001243103.1 | 2 - 3 | 213 | 70 | |
| GenBank mRNA | AA119196.1 | - | 2 - 3 | 190 | 70 | |
| AK080367.1 | - | 214 | 70 | |||
Set Membership: |
|