Search
Search

Odf3l2| Gene Symbol: Odf3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033473.2 | NP_001028645.1 | 2 - 3 | 279 | 70 | |
| GenBank mRNA | AK158063.1 | - | 2 - 3 | 280 | 70 | |
| BC147356.1 | - | 2 - 3 | 267 | 9056405 | 70 | |
| BC147357.1 | - | 2 - 3 | 267 | 9056398 | 70 | |
Set Membership: |
|