Search
Search
| Gene Symbol: Vax1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009501.1 | NP_033527.1 | 1 - 2 | 238 | 80 | |
| GenBank mRNA | AF064554.1 | - | 1 - 2 | 238 | 80 | |
| BC111818.1 | - | 1 - 2 | 237 | 40044698 | 80 | |
| BC119455.1 | - | 1 - 2 | 236 | 80 | ||
Set Membership: |
|