Search
Search

Zfp40| Gene Symbol: Zfp40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009555.2 | NP_033581.2 | 4 - 5 | 456 | 88 | |
| GenBank mRNA | AK049894.1 | - | 4 - 5 | 462 | 88 | |
| BC061205.1 | - | 4 - 5 | 425 | 30248495 | 88 | |
| BC065097.1 | - | 4 - 5 | 449 | 88 | ||
Set Membership: |
|