Search
Search

Krtap8-1| Gene Symbol: Krtap8-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010675.1 | NP_034805.1 | 1 | 121 | 97 | |
| GenBank mRNA | AK133727.1 | - | 1 | 122 | 97 | |
| D86423.1 | - | 1 | 54 | 97 | ||
Set Membership: |
|