Search
Search

Foxl2| Gene Symbol: Foxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012020.2 | NP_036150.1 | 1 | 919 | 109 | |
| GenBank mRNA | BC137811.1 | - | 1 | 919 | 8860830 | 109 |
| BC137812.1 | - | 1 | 919 | 8860831 | 109 | |
Set Membership: |
|