Search
Search

Lamtor3| Gene Symbol: Lamtor3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019920.2 | NP_064304.1 | 5 - 6 | 386 | 111 | |
| GenBank mRNA | AF082526.1 | - | 5 - 6 | 386 | 111 | |
| AK088454.1 | - | 5 - 6 | 393 | 111 | ||
| AK151432.1 | - | 5 - 6 | 416 | 111 | ||
| AK159450.1 | - | 5 - 6 | 384 | 111 | ||
| AK162102.1 | - | 5 - 6 | 378 | 111 | ||
| BC016417.1 | - | 5 - 6 | 369 | 3158151 | 111 | |
| BC083155.1 | - | 5 - 6 | 368 | 5339040 | 111 | |
Set Membership: |
|