Search
Search

Dnaja3| Gene Symbol: Dnaja3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135112.1 | NP_001128584.1 | 7 - 8 | 1061 | 62 | |
| NM_023646.4 | NP_076135.3 | 7 - 8 | 1061 | 62 | ||
| GenBank mRNA | AF325535.1 | - | 7 - 8 | 1102 | 62 | |
| AF326358.1 | - | 6 - 7 | 949 | 62 | ||
| AK004575.1 | - | 7 - 8 | 1011 | 62 | ||
| AK004910.1 | - | 7 - 8 | 1013 | 62 | ||
| AK031250.1 | - | 7 - 8 | 1018 | 62 | ||
| AK167511.1 | - | 7 - 8 | 1023 | 62 | ||
| AY009320.1 | - | 7 - 8 | 1102 | 62 | ||
| BC003920.1 | - | 7 - 8 | 994 | 62 | ||
| BC027240.1 | - | 7 - 8 | 1023 | 2654866 | 62 | |
Set Membership: |
|