Search
Search

Derl1| Gene Symbol: Derl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024207.4 | NP_077169.1 | 3 - 4 | 446 | 94 | |
| GenBank mRNA | AK007435.1 | - | 3 - 4 | 451 | 94 | |
| AK011499.1 | - | 3 - 4 | 468 | 94 | ||
| AK043420.1 | - | 3 - 4 | 447 | 94 | ||
| AK075631.1 | - | 3 - 4 | 347 | 94 | ||
| AK148018.1 | - | 3 - 4 | 436 | 94 | ||
| AK152197.1 | - | 3 - 4 | 444 | 94 | ||
| AK152901.1 | - | 3 - 4 | 426 | 94 | ||
| AK161702.1 | - | 3 - 4 | 473 | 95 | ||
| AK218964.1 | - | 3 - 4 | 217 | 94 | ||
| BC003454.1 | - | 3 - 4 | 471 | 2655640 | 94 | |
| BC085490.1 | - | 3 - 4 | 476 | 5715726 | 94 | |
Set Membership: |
|