Search
Search

Zfp763| Gene Symbol: Zfp763 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028543.3 | NP_082819.1 | 3 - 4 | 273 | 102 | |
| GenBank mRNA | AK006897.1 | - | 3 - 4 | 325 | 102 | |
| AK088750.1 | - | 3 - 4 | 102 | |||
| AK141711.1 | - | 3 - 4 | 274 | 102 | ||
| BC056193.1 | - | 3 - 4 | 260 | 30011543 | 102 | |
Set Membership: |
|