Search
Search
| Gene Symbol: Unkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197024.1 | NP_001183953.1 | 14 - 15 | 2037 | 76 | |
| NM_001290736.1 | NP_001277665.1 | 14 - 15 | 2048 | 76 | ||
| NM_028789.3 | NP_083065.1 | 4 - 5 | 676 | 76 | ||
| GenBank mRNA | AK004898.1 | - | 4 - 5 | 677 | 76 | |
| BC059910.1 | - | 14 - 15 | 2048 | 76 | ||
| BC089378.1 | - | 5 - 6 | 790 | 76 | ||
| BC150842.1 | - | 4 - 5 | 601 | 9087753 | 76 | |
Set Membership: |
|