Search
Search

Kbtbd13| Gene Symbol: Kbtbd13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028974.1 | NP_083250.1 | 1 | 1223 | 93 | |
| GenBank mRNA | AK017385.1 | - | 1 | 236 | 93 | |
| AK048611.1 | - | 1 | 1116 | 93 | ||
| BC139793.1 | - | 2 | 1103 | 93 | ||
Set Membership: |
|