Search
Search

4930513O06Rik| Gene Symbol: 4930513O06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029174.1 | NP_083450.1 | 2 - 3 | 517 | 148 | |
| GenBank mRNA | AK015788.1 | - | 2 - 3 | 518 | 148 | |
Set Membership: |
|