Search
Search

1700008F21Rik| Gene Symbol: 1700008F21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168369.1 | NP_001161841.1 | 7 - 8 | 877 | 86 | |
| NM_029292.3 | NP_083568.2 | 3 - 4 | 372 | 86 | ||
| GenBank mRNA | AK005764.1 | - | 3 - 4 | 373 | 86 | |
| AK076548.1 | - | 10 - 11 | 1150 | 86 | ||
| BC050962.1 | - | 6 - 7 | 399 | 86 | ||
| BC061058.1 | - | 4 - 5 | 276 | 86 | ||
| BC147560.1 | - | 7 - 8 | 882 | 9056550 | 86 | |
Set Membership: |
|