Search
Search

Olfr125| Gene Symbol: Olfr125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146290.2 | NP_666402.2 | 1 | 820 | 76 | |
| GenBank mRNA | BC116810.1 | - | 1 | 820 | 40126129 | 76 |
| BC116812.1 | - | 1 | 820 | 40126131 | 76 | |
Set Membership: |
|