Search
Search

Olfr12| Gene Symbol: Olfr12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206896.2 | NP_996779.1 | 1 | 1018 | 92 | |
| GenBank mRNA | BC120559.1 | - | 1 | 1018 | 40129482 | 92 |
| BC120561.1 | - | 1 | 1018 | 40129484 | 92 | |
Set Membership: |
|