Search
Search

Fam71a| Gene Symbol: Fam71a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109759.1 | NP_001103229.1 | 1 | 2016 | 72 | |
| GenBank mRNA | AB473728.1 | - | 1 | 1643 | 72 | |
| AK133412.1 | - | 1 | 2018 | 72 | ||
Set Membership: |
|