Search
Search

Ccdc90b| Gene Symbol: Ccdc90b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162918.1 | NP_001156390.1 | 5 - 6 | 826 | 71 | |
| NM_025515.3 | NP_079791.2 | 6 - 7 | 928 | 71 | ||
| GenBank mRNA | AK009370.1 | - | 6 - 7 | 549 | 71 | |
| AK084673.1 | - | 6 - 7 | 563 | 71 | ||
| AK160230.1 | - | 6 - 7 | 584 | 71 | ||
| BC034878.1 | - | 5 - 6 | 756 | 3468518 | 71 | |
| BC115973.1 | - | 6 - 7 | 549 | 40100643 | 71 | |
Set Membership: |
|