Search
Search

1700034J05Rik| Gene Symbol: 1700034J05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164236.1 | NP_001157708.1 | 2 - 3 | 831 | 71 | |
| GenBank mRNA | AK076643.1 | - | 2 - 3 | 825 | 71 | |
Set Membership: |
|