Search
Search

Trim6| Gene Symbol: Trim6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013616.2 | NP_001013637.1 | 2 - 3 | 562 | 88 | |
| GenBank mRNA | AF220031.1 | - | 1 - 2 | 260 | 88 | |
| AK033396.1 | - | 2 - 3 | 542 | 88 | ||
| AK040000.1 | - | 2 - 3 | 565 | 88 | ||
| AK049238.1 | - | 2 - 3 | 542 | 88 | ||
| BC138965.1 | - | 2 - 3 | 493 | 8861987 | 88 | |
Set Membership: |
|