Search
Search

Zfp961| Gene Symbol: Zfp961 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164581.1 | NP_001158053.1 | 1 - 2 | 100 | 59 | |
| GenBank mRNA | AK158413.1 | - | 1 - 2 | 102 | 59 | |
| BC049349.1 | - | 1 - 2 | 57 | 59 | ||
| BC065788.1 | - | 1 - 2 | 85 | 59 | ||
Set Membership: |
|