Search
Search

Klk1b16| Gene Symbol: Klk1b16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008454.2 | NP_032480.1 | 4 - 5 | 1179 | 128 | |
| GenBank mRNA | BC012237.1 | - | 4 - 5 | 650 | 4165443 | 128 |
| J03877.1 | - | 4 - 5 | 1180 | 128 | ||
Set Membership: |
|