Search
Search

H2-Q4| Gene Symbol: H2-Q4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143689.1 | NP_001137161.1 | 6 | 1420 | 99 | |
| GenBank mRNA | AK037574.1 | - | 6 | 1421 | 99 | |
| AK136497.1 | - | 1608 | 99 | |||
Set Membership: |
|