Search
Search

Armcx5| Gene Symbol: Armcx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009575.5 | NP_001009575.2 | 4 | 2121 | 108 | |
| GenBank mRNA | AK133950.1 | - | 4 | 2123 | 108 | |
| AK166624.1 | - | 5 | 1997 | 108 | ||
| BC027106.1 | - | 1 | 1154 | 5358405 | 108 | |
Set Membership: |
|