Search
Search

4833439L19Rik| Gene Symbol: 4833439L19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252645.1 | NP_001239574.1 | 7 | 907 | 82 | |
| NM_001252646.1 | NP_001239575.1 | 6 | 800 | 82 | ||
| NM_001252647.1 | NP_001239576.1 | 5 | 621 | 82 | ||
| NM_001252648.1 | NP_001239577.1 | 4 | 494 | 82 | ||
| NM_133797.4 | NP_598558.1 | 8 | 995 | 82 | ||
| GenBank mRNA | AK004845.1 | - | 8 | 954 | 82 | |
| AK014792.1 | - | 4 | 428 | 82 | ||
| AK028697.1 | - | 8 | 928 | 82 | ||
| AK145025.1 | - | 8 | 961 | 82 | ||
| AK159945.1 | - | 8 | 958 | 82 | ||
| AK167616.1 | - | 8 | 939 | 82 | ||
| AK169399.1 | - | 8 | 961 | 82 | ||
| AK173126.1 | - | 8 | 908 | 82 | ||
| BC033445.1 | - | 8 | 974 | 4222839 | 82 | |
| BC039775.1 | - | 8 | 933 | 4239147 | 82 | |
| BC043040.1 | - | 8 | 951 | 5701249 | 82 | |
| BC054417.1 | - | 8 | 940 | 3969623 | 82 | |
Set Membership: |
|