Search
Search

Zfp936| Gene Symbol: Zfp936 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034893.1 | NP_001030065.1 | 4 | 2717 | 83 | |
| GenBank mRNA | AK145615.1 | - | 4 | 2717 | 83 | |
| AK161338.1 | - | 1829 | 83 | |||
| BC066874.1 | - | 1 | 2647 | 30106557 | 83 | |
Set Membership: |
|