Search
Search

Tfb2m| Gene Symbol: Tfb2m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008249.4 | NP_032275.2 | 5 | 911 | 170 | |
| GenBank mRNA | AK146563.1 | - | 5 | 912 | 170 | |
| AK166717.1 | - | 5 | 898 | 170 | ||
| AK167930.1 | - | 5 | 863 | 170 | ||
| AK168970.1 | - | 5 | 890 | 170 | ||
| AK169138.1 | - | 5 | 890 | 170 | ||
| AK209110.1 | - | 1 | 51 | 225 | ||
| BC138836.1 | - | 5 | 856 | 8861858 | 170 | |
| BC138837.1 | - | 5 | 856 | 8861859 | 170 | |
| M74555.1 | - | 5 | 862 | 170 | ||
Set Membership: |
|