Search
Search

Iqcf5| Gene Symbol: Iqcf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029300.1 | NP_083576.1 | 2 | 423 | 94 | |
| GenBank mRNA | AK005734.1 | - | 2 | 424 | 94 | |
| BC125546.1 | - | 2 | 382 | 40130061 | 94 | |
Set Membership: |
|