Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAZAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012174256.2 | XP_012029646.1 | 3 - 4 | 471 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |