Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CCKBR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012095888.3 | XP_011951278.2 | 4 - 5 | 940 | 82 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Ovis aries |