Search
Search

CELF2| Gene Symbol: CELF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027976511.1 | XP_027832312.1 | 8 - 9 | 3351 | 62 | |
| XM_027976512.1 | XP_027832313.1 | 8 - 9 | 3351 | 62 | ||
| XM_027976513.1 | XP_027832314.1 | 8 - 9 | 3351 | 62 | ||
| XM_027976514.1 | XP_027832315.1 | 8 - 9 | 3350 | 62 | ||
| XM_027976515.1 | XP_027832316.1 | 8 - 9 | 3350 | 62 | ||
| XM_027976516.1 | XP_027832317.1 | 8 - 9 | 1972 | 62 | ||
| XM_027976517.1 | XP_027832318.1 | 8 - 9 | 1972 | 62 | ||
| XM_027976518.1 | XP_027832319.1 | 8 - 9 | 1971 | 62 | ||
| XM_027976519.1 | XP_027832320.1 | 9 - 10 | 975 | 62 | ||
| XM_027976520.1 | XP_027832321.1 | 8 - 9 | 864 | 62 | ||
| XM_027976521.1 | XP_027832322.1 | 8 - 9 | 864 | 62 | ||
| XM_027976522.1 | XP_027832323.1 | 8 - 9 | 864 | 62 | ||
| XM_027976523.1 | XP_027832324.1 | 8 - 9 | 864 | 62 | ||
| XM_027976524.1 | XP_027832325.1 | 8 - 9 | 816 | 62 | ||
| XM_027976526.1 | XP_027832327.1 | 6 - 7 | 679 | 62 | ||
| XM_027976527.1 | XP_027832328.1 | 8 - 9 | 1973 | 62 | ||
Set Membership: |
|