Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Oa04829224_m1
          See other PDE4DIP GE Assays ›
          Gene Symbol:
          PDE4DIP
          Gene Name:
          phosphodiesterase 4D interacting protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 102583352 - 102827976 on Build Oar_rambouillet_v1.0
          Species:
          Sheep
          Species Specific ID (Flybase ID):
          -
          Assay ID Oa04829224_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PDE4DIP
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_004002395.4 XP_004002444.2 10 - 11 2468 96
          XM_012179684.3 XP_012035074.2 10 - 11 2468 96
          XM_027973707.1 XP_027829508.1 10 - 11 2836 96
          XM_027973711.1 XP_027829512.1 10 - 11 2468 96
          XM_027973716.1 XP_027829517.1 17 - 18 2512 96
          XM_027973719.1 XP_027829520.1 17 - 18 2512 96
          XM_027973725.1 XP_027829526.1 10 - 11 2468 96
          XM_027973730.1 XP_027829531.1 10 - 11 2468 96
          XM_027973750.1 XP_027829551.1 10 - 11 2468 96
          XM_027973757.1 XP_027829558.1 10 - 11 2468 96
          XM_027973761.1 XP_027829562.1 17 - 18 2512 96
          XM_027973767.1 XP_027829568.1 10 - 11 2468 96
          XM_027973771.1 XP_027829572.1 10 - 11 2468 96
          XM_027973777.1 XP_027829578.1 17 - 18 2420 96
          XM_027973783.1 XP_027829584.1 17 - 18 2418 96
          XM_027973790.1 XP_027829591.1 10 - 11 2468 96
          XM_027973796.1 XP_027829597.1 17 - 18 2512 96
          XM_027973806.1 XP_027829607.1 17 - 18 2512 96
          XM_027973821.1 XP_027829622.1 14 - 15 2164 96
          XM_027973827.1 XP_027829628.1 14 - 15 2143 96
          XM_027973834.1 XP_027829635.1 14 - 15 2164 96
          XM_027973837.1 XP_027829638.1 14 - 15 2164 96
          XM_027973847.1 XP_027829648.1 17 - 18 2512 96
          XM_027973848.1 XP_027829649.1 17 - 18 2512 96
          XM_027973850.1 XP_027829651.1 17 - 18 2512 96

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 Ovis aries

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline