Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082734.1 | NP_001076203.1 | 2 - 3 | 417 | 80 | |
XM_008258167.2 | XP_008256389.1 | 3 - 4 | 534 | 80 | ||
GenBank mRNA | AY225335.1 | - | 2 - 3 | 417 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Oryctolagus cuniculus Probe spans exons |