Search
Search

AOX2| Gene Symbol: AOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297488.1 | NP_001284417.1 | 20 - 21 | 2229 | 65 | |
| XM_017342642.1 | XP_017198131.1 | 20 - 21 | 2373 | 65 | ||
Set Membership: |
|