Search
Search

LOC4347953| Gene Symbol: LOC4347953 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759291.2 | XP_015614777.1 | 1 - 2 | 101 | 150 | |
| GenBank mRNA | AK071821.1 | - | 1 - 2 | 102 | 150 | |
| AK100339.1 | - | 1 - 2 | 102 | 150 | ||
Set Membership: |
|