Search
Search

LOC4327109| Gene Symbol: LOC4327109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755428.2 | XP_015610914.1 | 4 - 5 | 551 | 68 | |
| GenBank mRNA | AK071420.1 | - | 4 - 5 | 474 | 68 | |
Set Membership: |
|