Search
Search

GPX1| Gene Symbol: GPX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159298.2 | NP_001152770.1 | 315 | 74 | ||
| GenBank mRNA | DR771003.1 | - | 319 | 74 | ||
| JU485035.1 | - | 307 | 74 | |||
| JV503934.1 | - | 176 | 74 | |||
| JV614160.1 | - | 314 | 74 | |||
Set Membership: |
|